Assay Details
Gene Symbol: Olfr1220 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146900.2 | NP_667111.2 | 1 | 879 | 120 | |
GenBank mRNA | BC125363.1 | - | 1 | 879 | 40129878 | 120 |
BC131900.1 | - | 1 | 879 | 40130177 | 120 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2017129 | BC131900.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2017129 | BC125363.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2017129 | BC131900.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2017129 | BC125363.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P322781 | BC131900.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P322781 | NM_146900.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P322781 | BC125363.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P322781 | BC131900.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P322781 | NM_146900.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P322781 | BC125363.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P137636 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412477 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412453 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412486 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412522 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412452 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412518 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412513 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412474 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412505 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412532 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412450 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412488 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412511 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412491 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412524 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412479 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412539 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412509 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412497 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412481 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412533 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412503 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412534 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412482 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412493 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412527 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412535 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412460 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412446 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412538 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412489 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412499 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412536 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412458 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412492 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412466 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412529 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412537 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412454 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412517 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412448 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412530 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412484 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21412519 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21412500 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21412491 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01486104 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21412486 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21412459 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21412539 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21412499 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21412528 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21412530 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01923008 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21412511 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21412460 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21412482 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21412536 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21412458 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21412477 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21412489 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21412517 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21412474 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21412521 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21412492 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21412484 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21412488 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21412481 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21412462 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21412494 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21412538 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21412518 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21412479 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21412524 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21412457 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21412503 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21412534 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21412535 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21412497 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21412532 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21412519 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21412507 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21412493 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21412505 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21412533 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02080462 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21412522 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21412509 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21412527 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01947174 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21412513 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21412516 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21412529 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10484787 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10484788 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10484786 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10484785 | - | - |
Affymetrix Mouse Clariom S Array | PSR0200034003.mm.1 | NM_146900.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200034003.mm.1 | NM_146900.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200034003.mm.1 | TC0200004040.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 472256 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 6620671 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006620671 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2674041 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2674041 | - | - |
mouse-6 v1.1 (Illumina) | scl4293.1.1_235-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6620671 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl4293.1.1_235-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6620671 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2798069 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2798069 | - | - |
