Assay Details
| Gene Symbol: Cd59a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111060.2 | NP_001104530.1 | 1 - 2 | 123 | 83 | |
| GenBank mRNA | CA467821.1 | - | 121 | 83 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001111060.2 transcripts.
Set Membership: |
|

Cd59a
Genomic Map