| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200011661.mm.1 |
NM_001111060.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200011661.mm.1 |
NM_007652.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200011661.mm.1 |
TC0200001552.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200011660.mm.1 |
NM_001111060.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200011660.mm.1 |
NM_007652.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200011660.mm.1 |
TC0200001552.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200011664.mm.1 |
NM_001111060.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200011664.mm.1 |
NM_007652.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200011664.mm.1 |
TC0200001552.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200011662.mm.1 |
NM_001111060.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200011662.mm.1 |
TC0200001552.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200011665.mm.1 |
NM_001111060.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200011665.mm.1 |
NM_007652.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200011665.mm.1 |
TC0200001552.mm.1 |
MTA TC |