| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700016004.mm.1 |
TC0700001861.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700016004.mm.1 |
NM_172748.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700016004.mm.1 |
NM_172748.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700016005.mm.1 |
TC0700001861.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700016005.mm.1 |
NM_172748.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700016005.mm.1 |
NM_172748.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700016003.mm.1 |
TC0700001861.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700016003.mm.1 |
NM_172748.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700016003.mm.1 |
NM_172748.2 |
Refseq |