Assay Details
| Gene Symbol: Fuom | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286217.1 | NP_001273146.1 | 2 - 3 | 200 | 55 | |
| NM_026928.3 | NP_081204.2 | 2 - 3 | 200 | 55 | ||
| NR_104415.1 | 2 - 3 | 200 | 55 | |||
| GenBank mRNA | AK007494.1 | - | 2 - 3 | 163 | 55 | |
| AK042452.1 | - | 163 | 55 | |||
| AK044731.1 | - | 2 - 3 | 165 | 55 | ||
| AK079410.1 | - | 2 - 3 | 166 | 55 | ||
| AK163708.1 | - | 2 - 3 | 104 | 55 | ||
| BC028662.1 | - | 2 - 3 | 161 | 1332340 | 55 | |
| CK618091.1 | - | 2 - 3 | 135 | 55 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fuom
Genomic Map