Assay Details
| Gene Symbol: Prss53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081268.1 | NP_001074737.1 | 1 - 2 | 108 | 124 | |
| GenBank mRNA | AK220244.1 | - | 1 - 2 | 65 | 124 | |
| BC147509.1 | - | 1 - 2 | 105 | 9056457 | 124 | |
| BC151010.1 | - | 1 - 2 | 105 | 9087923 | 124 | |
| BC151017.1 | - | 1 - 2 | 105 | 9087930 | 124 | |
| BC172059.1 | - | 1 - 2 | 105 | 9054345 | 124 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prss53
Genomic Map