Assay Details
| Gene Symbol: Prss53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081268.1 | NP_001074737.1 | 7 - 8 | 1087 | 60 | |
| GenBank mRNA | AK220244.1 | - | 7 - 8 | 1044 | 60 | |
| BC039632.1 | - | 4 - 5 | 657 | 60 | ||
| BC147509.1 | - | 7 - 8 | 1084 | 9056457 | 60 | |
| BC151010.1 | - | 7 - 8 | 1084 | 9087923 | 60 | |
| BC151017.1 | - | 7 - 8 | 1084 | 9087930 | 60 | |
| BC172059.1 | - | 7 - 8 | 1084 | 9054345 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prss53
Genomic Map