Assay Details
| Gene Symbol: Prss53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081268.1 | NP_001074737.1 | 9 - 10 | 1475 | 90 | |
| GenBank mRNA | AK220244.1 | - | 9 - 10 | 1432 | 90 | |
| BC039632.1 | - | 6 - 7 | 1045 | 90 | ||
| BC147509.1 | - | 9 - 10 | 1472 | 9056457 | 90 | |
| BC151010.1 | - | 9 - 10 | 1472 | 9087923 | 90 | |
| BC151017.1 | - | 9 - 10 | 1472 | 9087930 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2064841 | NM_001081268.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2064841 | AK220244.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2064841 | NM_172749.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2064841 | AK078520.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2064841 | AK129112.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 633981 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 730898 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 671593 | AK089249.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 671593 | AK089470.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 671593 | NM_010705.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 24939 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P161297 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2064841 | NM_001081268.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2064841 | AK220244.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2064841 | NM_172749.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2064841 | AK078520.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2064841 | AK129112.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P501167 | NM_001081268.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P501167 | AK220244.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P501167 | BC039632.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25507834 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507945 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507916 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507919 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507949 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06290993 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507917 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06095853 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06290960 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06291010 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507910 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507923 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507909 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06290998 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507928 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507926 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507907 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507940 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06284996 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507935 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507950 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507929 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507952 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507953 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507951 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06290985 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507948 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06290953 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507915 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06284997 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507937 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507905 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507925 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507902 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507908 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507898 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06290980 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507939 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25507954 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10568324 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568321 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568327 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568319 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568323 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568320 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568322 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568326 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568325 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1441991_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10568318 | - | - |
| Mouse Genome 430 2.0 Array | 1441991_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1441991_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 330657_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 330657_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700036491.mm.1 | NM_001081268.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700036491.mm.1 | NM_001081268.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700036491.mm.1 | TC0700004664.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700036494.mm.1 | NM_001081268.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700036494.mm.1 | NM_001081268.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700036494.mm.1 | TC0700004664.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700039200.mm.1 | NM_001081268.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700039200.mm.1 | NM_001081268.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700039200.mm.1 | TC0700004664.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700036490.mm.1 | NM_001081268.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700036490.mm.1 | NM_001081268.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700036490.mm.1 | TC0700004664.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1441991_at | - | - |
| Mouse Expression Set 430 | 1441991_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1441991_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1441991_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 486235 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2706460 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2682674 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2682674 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2706460 | - | - |

Prss53
Genomic Map