Assay Details
| Gene Symbol: Urah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029821.2 | NP_084097.1 | 2 - 3 | 394 | 91 | |
| GenBank mRNA | AK004470.1 | - | 2 - 3 | 612 | 91 | |
| AK013117.1 | - | 2 - 3 | 396 | 91 | ||
| BC051545.1 | - | 2 - 3 | 562 | 5255010 | 91 | |
| DQ225768.1 | - | 2 - 3 | 269 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2174582 | BC051545.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2174582 | NM_029821.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2174582 | AK004470.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2174582 | AK013117.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1971006 | BC051545.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1971006 | NM_029821.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1971006 | AK013117.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 345966 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P171619 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2174582 | BC051545.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2174582 | NM_029821.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2174582 | AK004470.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2174582 | AK013117.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1971006 | BC051545.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1971006 | NM_029821.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1971006 | AK013117.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P171692 | AK004470.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P389479 | AK004470.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P171619 | - | - |
| Whole Mouse Genome G4122A | A_51_P389479 | AK004470.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P171692 | AK004470.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P171619 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06300605 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06300583 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06300600 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06300598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25585184 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585099 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585113 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585136 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06300588 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06357779 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585132 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585098 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585128 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585125 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585121 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585110 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585130 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585106 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585134 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06300600 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585101 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585115 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585119 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585094 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06585082 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585096 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25585103 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06300579 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06300571 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10558692 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558690 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558689 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558691 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558688 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558693 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455186_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1431786_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10558687 | - | - |
| Mouse Genome 430 2.0 Array | 1431786_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1431786_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1455186_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1455186_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 76974_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 76974_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700017722.mm.1 | TC0700002024.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700017722.mm.1 | NM_029821.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700017722.mm.1 | NM_029821.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700017723.mm.1 | TC0700002024.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700017723.mm.1 | NM_029821.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700017723.mm.1 | NM_029821.2 | Refseq |
| Mouse Expression Set 430 | 1455186_PM_a_at | - | - |
| Mouse Expression Set 430 | 1455186_a_at | - | - |
| Mouse Expression Set 430 | 1431786_PM_s_at | - | - |
| Mouse Expression Set 430 | 1431786_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431786_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455186_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431786_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455186_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 908644 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2734924 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2734924 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2734924 | - | - |

Urah
Genomic Map