Assay Details
| Gene Symbol: Slc24a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053195.2 | NP_444425.1 | 9 - 10 | 673 | 88 | |
| GenBank mRNA | AF314821.1 | - | 9 - 10 | 713 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc24a3
Genomic Map