Assay Details
| Gene Symbol: Ppfia1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033319.2 | NP_001028491.1 | 19 - 20 | 2728 | 74 | |
| NM_001195086.1 | NP_001182015.1 | 20 - 21 | 2803 | 74 | ||
| GenBank mRNA | BC038349.1 | - | 8 - 9 | 1091 | 74 | |
| BC157936.1 | - | 20 - 21 | 2620 | 9088116 | 74 | |
| BC158007.1 | - | 19 - 20 | 2685 | 9007315 | 74 | |
| BC172096.1 | - | 20 - 21 | 2623 | 9054382 | 74 | |
| EU568869.1 | - | 20 - 21 | 2803 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppfia1
Genomic Map