Assay Details
| Gene Symbol: Ppfia1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033319.2 | NP_001028491.1 | 21 - 22 | 2899 | 68 | |
| NM_001195086.1 | NP_001182015.1 | 22 - 23 | 2974 | 68 | ||
| GenBank mRNA | BC038349.1 | - | 10 - 11 | 1262 | 68 | |
| BC157936.1 | - | 22 - 23 | 2791 | 9088116 | 68 | |
| BC158007.1 | - | 21 - 22 | 2856 | 9007315 | 68 | |
| BC172096.1 | - | 22 - 23 | 2794 | 9054382 | 68 | |
| EU568869.1 | - | 22 - 23 | 2974 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppfia1
Genomic Map