Assay Details
| Gene Symbol: Ppfia1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033319.2 | NP_001028491.1 | 26 - 27 | 3471 | 121 | |
| NM_001195086.1 | NP_001182015.1 | 27 - 28 | 3546 | 121 | ||
| GenBank mRNA | AK146526.1 | - | 3 - 4 | 420 | 121 | |
| BC038349.1 | - | 13 - 14 | 1744 | 121 | ||
| BC157936.1 | - | 27 - 28 | 3363 | 9088116 | 121 | |
| BC158007.1 | - | 26 - 27 | 3428 | 9007315 | 121 | |
| BC172096.1 | - | 27 - 28 | 3366 | 9054382 | 121 | |
| EU568869.1 | - | 27 - 28 | 3549 | 121 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppfia1
Genomic Map