Assay Details
| Gene Symbol: Ppfia1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033319.2 | NP_001028491.1 | 3 - 4 | 573 | 66 | |
| NM_001195086.1 | NP_001182015.1 | 3 - 4 | 573 | 66 | ||
| GenBank mRNA | BC157936.1 | - | 3 - 4 | 390 | 9088116 | 66 |
| BC158007.1 | - | 3 - 4 | 530 | 9007315 | 66 | |
| BC172096.1 | - | 3 - 4 | 393 | 9054382 | 66 | |
| EU568869.1 | - | 3 - 4 | 573 | 66 | ||
More Information
Set Membership: |
|

Ppfia1
Genomic Map