Assay Details
| Gene Symbol: Wipf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289722.1 | NP_001276651.1 | 5 - 6 | 1304 | 82 | |
| NM_001289723.1 | NP_001276652.1 | 5 - 6 | 1269 | 82 | ||
| NM_153138.4 | NP_694778.1 | 5 - 6 | 1299 | 82 | ||
| GenBank mRNA | AJ437262.1 | - | 4 - 5 | 1100 | 82 | |
| AK031046.1 | - | 5 - 6 | 1266 | 82 | ||
| AK156547.1 | - | 5 - 6 | 1304 | 82 | ||
| AK168092.1 | - | 1 - 2 | 453 | 82 | ||
| BC049788.1 | - | 5 - 6 | 1222 | 6514406 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wipf1
Genomic Map