| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200032221.mm.1 |
NM_001025093.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200032221.mm.1 |
NM_001284369.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200032221.mm.1 |
NM_001284370.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200032221.mm.1 |
NM_001284371.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200032221.mm.1 |
NM_001284372.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200032221.mm.1 |
NM_001284373.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200032221.mm.1 |
NM_001284374.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200032221.mm.1 |
NM_001284376.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200032221.mm.1 |
NM_009715.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200032221.mm.1 |
TC0200003763.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200032222.mm.1 |
NM_001025093.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200032222.mm.1 |
NM_001284369.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200032222.mm.1 |
NM_001284370.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200032222.mm.1 |
NM_001284371.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200032222.mm.1 |
NM_001284372.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200032222.mm.1 |
NM_001284373.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200032222.mm.1 |
NM_001284374.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200032222.mm.1 |
NM_001284376.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200032222.mm.1 |
NM_009715.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200032222.mm.1 |
TC0200003763.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200032216.mm.1 |
NM_001025093.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200032216.mm.1 |
NM_001284369.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200032216.mm.1 |
NM_001284370.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200032216.mm.1 |
NM_001284371.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200032216.mm.1 |
NM_001284372.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200032216.mm.1 |
NM_001284373.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200032216.mm.1 |
NM_001284374.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200032216.mm.1 |
NM_001284376.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200032216.mm.1 |
NM_009715.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200032216.mm.1 |
TC0200003763.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200032229.mm.1 |
NM_001025093.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200032229.mm.1 |
NM_001284369.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200032229.mm.1 |
NM_001284370.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200032229.mm.1 |
NM_001284371.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200032229.mm.1 |
NM_001284372.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200032229.mm.1 |
NM_001284373.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200032229.mm.1 |
NM_001284374.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200032229.mm.1 |
NM_001284376.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200032229.mm.1 |
NM_009715.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200032229.mm.1 |
TC0200003763.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200032213.mm.1 |
NM_001025093.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200032213.mm.1 |
NM_001284370.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200032213.mm.1 |
NM_001284371.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200032213.mm.1 |
NM_001284372.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200032213.mm.1 |
NM_001284373.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200032213.mm.1 |
TC0200003763.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200032218.mm.1 |
NM_001025093.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200032218.mm.1 |
NM_001284370.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200032218.mm.1 |
NM_001284372.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200032218.mm.1 |
NM_001284376.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200032218.mm.1 |
NM_009715.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200032218.mm.1 |
TC0200003763.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200032224.mm.1 |
NM_001025093.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200032224.mm.1 |
NM_001284369.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200032224.mm.1 |
NM_001284370.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200032224.mm.1 |
NM_001284371.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200032224.mm.1 |
NM_001284372.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200032224.mm.1 |
NM_001284373.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200032224.mm.1 |
NM_001284374.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200032224.mm.1 |
NM_001284376.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200032224.mm.1 |
NM_009715.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200032224.mm.1 |
TC0200003763.mm.1 |
MTA TC |