Assay Details
| Gene Symbol: Myef2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162418.1 | NP_001155890.1 | 12 - 13 | 1369 | 53 | |
| GenBank mRNA | AK012133.1 | - | 12 - 13 | 1371 | 53 | |
| AK129336.1 | - | 13 - 14 | 1332 | 53 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Myef2
Genomic Map