Assay Details
| Gene Symbol: B4galnt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177897.3 | NP_808565.2 | 10 - 11 | 986 | 60 | |
| GenBank mRNA | AB114827.1 | - | 10 - 11 | 986 | 60 | |
| BC031982.1 | - | 9 - 10 | 840 | 60 | ||
| BC062653.1 | - | 6 - 7 | 676 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

B4galnt4
Genomic Map