Assay Details
| Gene Symbol: B4galnt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177897.3 | NP_808565.2 | 12 - 13 | 1198 | 133 | |
| GenBank mRNA | AB114827.1 | - | 12 - 13 | 1198 | 133 | |
| BC031982.1 | - | 11 - 12 | 1052 | 133 | ||
| BC062653.1 | - | 8 - 9 | 888 | 133 | ||
| BC079533.1 | - | 1 - 2 | 101 | 133 | ||
More Information
Updated Mapping Information
Set Membership: |
|

B4galnt4
Genomic Map