| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200033014.mm.1 |
NM_001113399.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200033014.mm.1 |
NM_001113399.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200033014.mm.1 |
NM_001113400.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200033014.mm.1 |
NM_001113400.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200033014.mm.1 |
NM_178723.6.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200033014.mm.1 |
NM_178723.6 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200033014.mm.1 |
TC0200003815.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200033021.mm.1 |
NM_001113400.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200033021.mm.1 |
NM_001113400.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200033021.mm.1 |
TC0200003815.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200033023.mm.1 |
NM_001113400.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200033023.mm.1 |
NM_001113400.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200033023.mm.1 |
TC0200003815.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200033020.mm.1 |
NM_001113400.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200033020.mm.1 |
NM_001113400.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200033020.mm.1 |
TC0200003815.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200033013.mm.1 |
NM_001113399.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200033013.mm.1 |
NM_001113399.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200033013.mm.1 |
NM_001113400.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200033013.mm.1 |
NM_001113400.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200033013.mm.1 |
NM_178723.6.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200033013.mm.1 |
NM_178723.6 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200033013.mm.1 |
TC0200003815.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200033016.mm.1 |
NM_001113399.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200033016.mm.1 |
NM_001113399.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200033016.mm.1 |
NM_001113400.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200033016.mm.1 |
NM_001113400.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200033016.mm.1 |
NM_178723.6.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200033016.mm.1 |
NM_178723.6 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200033016.mm.1 |
TC0200003815.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200033028.mm.1 |
NM_001113399.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200033028.mm.1 |
NM_001113399.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200033028.mm.1 |
TC0200003815.mm.1 |
MTA TC |