| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200033031.mm.1 |
NM_178723.6.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200033031.mm.1 |
NM_178723.6 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200033031.mm.1 |
TC0200003815.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200033034.mm.1 |
NM_178723.6.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200033034.mm.1 |
NM_178723.6 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200033034.mm.1 |
TC0200003815.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200033035.mm.1 |
NM_178723.6.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200033035.mm.1 |
NM_178723.6 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200033035.mm.1 |
TC0200003815.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200033032.mm.1 |
NM_178723.6.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200033032.mm.1 |
NM_178723.6 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200033032.mm.1 |
TC0200003815.mm.1 |
MTA TC |