Assay Details
| Gene Symbol: Tap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011530.3 | NP_035660.3 | 1 - 2 | 148 | 87 | |
| GenBank mRNA | AK020022.1 | - | 1 - 2 | 61 | 87 | |
| AK079667.1 | - | 1 - 2 | 61 | 87 | ||
| AK145234.1 | - | 1 - 2 | 105 | 87 | ||
| AK151376.1 | - | 1 - 2 | 61 | 87 | ||
| AK151483.1 | - | 1 - 2 | 61 | 87 | ||
| AK151657.1 | - | 1 - 2 | 81 | 87 | ||
| AK153287.1 | - | 1 - 2 | 61 | 87 | ||
| AK159200.1 | - | 1 - 2 | 114 | 87 | ||
| AK161091.1 | - | 1 - 2 | 62 | 87 | ||
| BC051257.1 | - | 1 - 2 | 61 | 6530488 | 87 | |
| BC057668.1 | - | 1 - 2 | 434 | 87 | ||
| CJ046345.1 | - | 1 - 2 | 149 | 87 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK151376.1,AK151483.1,AK153287.1,AK159200.1,BC051257.1,NM_011530.3 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tap2
Genomic Map