Assay Details
| Gene Symbol: Tbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013684.3 | NP_038712.3 | 3 - 4 | 673 | 65 | |
| GenBank mRNA | AK085037.1 | - | 3 - 4 | 674 | 65 | |
| BC012685.1 | - | 5 - 6 | 909 | 3498542 | 65 | |
| BC016476.1 | - | 3 - 4 | 651 | 3584341 | 65 | |
| D01034.1 | - | 2 - 3 | 490 | 65 | ||
| U63933.1 | - | 4 - 5 | 679 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tbp
Genomic Map