Assay Details
| Gene Symbol: Plekha7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC084587.1 | - | 1 - 2 | 148 | 3602481 | 59 |
| BY729140.1 | - | 1 - 2 | 196 | 59 | ||
| CD538709.1 | - | 1 - 2 | 258 | 59 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC084587.1 transcripts.
Set Membership: |
|

Plekha7
Genomic Map