Assay Details
| Gene Symbol: Trp53i11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025246.1 | NP_001020417.1 | 2 - 3 | 240 | 61 | |
| GenBank mRNA | AK142316.1 | - | 3 - 4 | 413 | 61 | |
| BC098206.1 | - | 2 - 3 | 240 | 6812412 | 61 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK142316.1, BC098206.1, NM_001025246.1 transcripts.
Set Membership: |
|

Trp53i11
Genomic Map