Assay Details
| Gene Symbol: Galk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291002.1 | NP_001277931.1 | 1 - 2 | 159 | 123 | |
| GenBank mRNA | AK053002.1 | - | 1 - 2 | 115 | 123 | |
| AK154762.1 | - | 1 - 2 | 114 | 123 | ||
| AK171939.1 | - | 1 - 2 | 142 | 123 | ||
| BC079843.1 | - | 1 - 2 | 129 | 5690056 | 123 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK053002.1, AK154762.1, BC079843.1, NM_001291002.1 transcripts.
Set Membership: |
|

Galk2
Genomic Map