Assay Details
| Gene Symbol: Galk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291002.1 | NP_001277931.1 | 4 - 5 | 464 | 100 | |
| NM_175154.3 | NP_780363.1 | 4 - 5 | 570 | 100 | ||
| GenBank mRNA | AK053002.1 | - | 4 - 5 | 420 | 100 | |
| AK082521.1 | - | 4 - 5 | 461 | 100 | ||
| AK154762.1 | - | 4 - 5 | 419 | 100 | ||
| AK171939.1 | - | 4 - 5 | 447 | 100 | ||
| AK176923.1 | - | 4 - 5 | 100 | |||
| BC079843.1 | - | 4 - 5 | 434 | 5690056 | 100 | |
More Information
Set Membership: |
|

Galk2
Genomic Map