Assay Details
| Gene Symbol: Cyp39a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285947.1 | NP_001272876.1 | 3 - 4 | 728 | 75 | |
| GenBank mRNA | BC037663.1 | - | 3 - 4 | 577 | 5352740 | 75 |
More Information
Updated Mapping Information
Set Membership: |
|

Cyp39a1
Genomic Map