Assay Details
| Gene Symbol: Rpp21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026308.2 | NP_080584.1 | 2 - 3 | 167 | 86 | |
| GenBank mRNA | AK002780.1 | - | 2 - 3 | 168 | 86 | |
| AK003455.1 | - | 2 - 3 | 165 | 86 | ||
| AY251016.1 | - | 2 - 3 | 189 | 86 | ||
| BC028491.1 | - | 2 - 3 | 177 | 1497187 | 86 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rpp21
Genomic Map