Assay Details
| Gene Symbol: Plcl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013880.3 | NP_038908.2 | 1 - 2 | 393 | 56 | |
| GenBank mRNA | AK154288.1 | - | 1 - 2 | 349 | 56 | |
| AK165548.1 | - | 1 - 2 | 395 | 56 | ||
| BC027746.1 | - | 1 - 2 | 455 | 5065782 | 56 | |
| BY125070.1 | - | 1 - 2 | 183 | 56 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Plcl2
Genomic Map