Assay Details
| Gene Symbol: Prrt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102563.1 | NP_001096033.1 | 2 - 3 | 1170 | 74 | |
| GenBank mRNA | AK005319.1 | - | 1 - 2 | 159 | 74 | |
| BC053419.1 | - | 3 - 4 | 686 | 74 | ||
| BF323287.1 | - | 1 - 2 | 450 | 74 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Prrt2
Genomic Map