Assay Details
| Gene Symbol: Acer3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025408.2 | NP_079684.2 | 6 - 7 | 509 | 193 | |
| GenBank mRNA | AK004287.1 | - | 6 - 7 | 510 | 193 | |
| AK011668.1 | - | 6 - 7 | 490 | 193 | ||
| AK017361.1 | - | 414 | 193 | |||
| AK080977.1 | - | 6 - 7 | 487 | 193 | ||
| AK150629.1 | - | 6 - 7 | 470 | 193 | ||
| AK162403.1 | - | 6 - 7 | 525 | 193 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Acer3
Genomic Map