Assay Details
| Gene Symbol: Acer3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025408.2 | NP_079684.2 | 7 - 8 | 571 | 98 | |
| GenBank mRNA | AK004287.1 | - | 7 - 8 | 572 | 98 | |
| AK011668.1 | - | 7 - 8 | 552 | 98 | ||
| AK017361.1 | - | 476 | 98 | |||
| AK080977.1 | - | 7 - 8 | 549 | 98 | ||
| AK150629.1 | - | 7 - 8 | 532 | 98 | ||
| AK162403.1 | - | 7 - 8 | 587 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Acer3
Genomic Map