Assay Details
| Gene Symbol: Stk33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_054103.1 | NP_473444.1 | 3 - 4 | 687 | 148 | |
| GenBank mRNA | AK014819.1 | - | 3 - 4 | 688 | 148 | |
More Information
Updated Mapping Information
Set Membership: |
|

Stk33
Genomic Map