Assay Details
| Gene Symbol: Stk33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_054103.1 | NP_473444.1 | 8 - 9 | 1244 | 117 | |
| GenBank mRNA | AK014819.1 | - | 8 - 9 | 1244 | 117 | |
| AM056057.1 | - | 5 - 6 | 512 | 117 | ||
| BC117798.1 | - | 5 - 6 | 533 | 117 | ||
| BC117799.1 | - | 5 - 6 | 533 | 117 | ||
| BC127158.1 | - | 5 - 6 | 533 | 117 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stk33
Genomic Map