Assay Details
| Gene Symbol: Tmco5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026104.4 | NP_080380.1 | 10 - 11 | 763 | 99 | |
| GenBank mRNA | AK007078.1 | - | 10 - 11 | 747 | 99 | |
| BC049568.1 | - | 10 - 11 | 727 | 6742928 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmco5
Genomic Map