Assay Details
| Gene Symbol: 1700007K09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027037.2 | NP_081313.1 | 1 - 2 | 65 | 149 | |
| GenBank mRNA | AK005730.1 | - | 1 - 2 | 60 | 149 | |
| BC048596.1 | - | 1 - 2 | 65 | 6740370 | 149 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK005730.1, BC048596.1, NM_027037.2 transcripts.
Set Membership: |
|

1700007K09Rik
Genomic Map