Assay Details
| Gene Symbol: Gm6249 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_046021.1 | 5 - 6 | 1259 | 63 | ||
| GenBank mRNA | AK034107.1 | - | 5 - 6 | 1260 | 63 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA C230079O03 gene | C230079O03Rik | C230079O03 | -318.5kb | Sense | Chr. 7 - 136332216 - 136349415 on NCBI Build GRCm38 |
| Downstream | O-6-methylguanine-DNA methyltransferase | Mgmt |
AGT, AI267024, Agat |
208.7kb | Anti-sense | Chr. 7 - 136894468 - 137128193 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2121761 | AK034107.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 113341 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 75477 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 190938 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 662623 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 112120 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 500178 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 664630 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 277622 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 895400 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 966320 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 177466 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 278770 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 7424 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 459374 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 166243 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 137943 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2121761 | AK034107.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06385106 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06385145 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133198 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06385145 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560793 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560836 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05953227 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560826 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06508330 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560728 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560789 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560734 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25561079 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560799 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560939 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06577148 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560923 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25561086 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560855 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560933 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560802 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560837 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560841 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560828 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06207326 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560792 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25561036 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560700 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560915 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560819 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06577159 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560737 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560924 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560854 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25561033 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06546076 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560931 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560721 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25561074 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560936 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560817 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560714 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560921 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560726 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560820 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560710 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25561085 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560944 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06567747 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25561077 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560825 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560705 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560832 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06613722 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25561039 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560857 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05977483 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560940 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560716 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560843 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560926 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25561042 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560813 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560835 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560719 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25561027 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06595685 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560919 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06207331 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06207323 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25561078 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560846 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06207311 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560928 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560913 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06287200 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560848 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25561037 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560704 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560804 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560712 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560724 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560822 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560938 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06207312 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560735 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25561043 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06508329 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25561031 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06613715 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560808 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25561035 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25561038 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560788 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06508321 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06197824 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05977476 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25561073 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560810 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25561088 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560795 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560920 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560932 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25561076 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25561080 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06567731 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560703 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560943 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25561041 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560853 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560708 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06207307 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560840 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25561028 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560797 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560917 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25560942 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Clariom S Array | PSR0700037420.mm.1 | NR_046021.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700037420.mm.1 | NR_046021.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700037420.mm.1 | TC0700004459.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 661200 | - | - |

Gm6249
Genomic Map