Assay Details
| Gene Symbol: Gm15506 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK052909.1 | - | 1 - 2 | 41 | 73 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 2210018M11 gene | 2210018M11Rik |
2310016L09Rik, AU019521, AU019557, Emsy |
-35.1kb | Sense | Chr. 7 - 98587137 - 98658289 on NCBI Build GRCm38 |
| Downstream |
protein-kinase, interferon-inducible double stranded RNA dependent inhibitor, repressor of (P58 repressor) |
Prkrir |
2900052B10Rik, Dap4, Rpkrir |
302.0 | Anti-sense | Chr. 7 - 98703230 - 98718062 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm15506
Genomic Map