Assay Details
Gene Symbol: H2-M5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115075.1 | NP_001108547.1 | 4 - 5 | 898 | 73 | |
GenBank mRNA | AB359228.2 | - | 4 - 5 | 898 | 73 | |
AK051129.1 | - | 1309 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2135079 | AK051129.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2135074 | NM_001115075.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2135074 | AB359228.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 362462 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 583264 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P115438 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2135079 | AK051129.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2135074 | NM_001115075.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2135074 | AB359228.2 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P115438 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P115438 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166704 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166592 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166662 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166657 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166701 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166651 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166594 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166700 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166647 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166604 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166631 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166683 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166706 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166666 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166618 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166682 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166678 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166648 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166667 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166664 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166617 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166646 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166681 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166707 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166624 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166702 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166692 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166611 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166591 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166649 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166621 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166614 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166695 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166590 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166677 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166653 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166703 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166596 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166625 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166629 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166698 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166684 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166689 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166658 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166687 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166607 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166660 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166674 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166655 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166669 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166633 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166627 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166588 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166601 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31166681 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13305115 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31166646 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31166665 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31166662 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31166669 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31166647 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31166655 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13578969 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31166664 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31166674 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31166660 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31166649 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31166651 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31166676 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13450539 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31166678 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13305110 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31166679 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31166677 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31166648 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31166687 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31166653 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31166683 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13428453 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31166657 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10450844 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10450843 | - | - |
Affymetrix Mouse Clariom S Array | PSR1700019495.mm.1 | NM_001115075.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700019495.mm.1 | NM_001115075.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700019495.mm.1 | TC1700002046.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700019499.mm.1 | NM_001115075.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700019499.mm.1 | NM_001115075.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700019499.mm.1 | TC1700002046.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700019502.mm.1 | NM_001115075.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700019502.mm.1 | NM_001115075.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700019502.mm.1 | TC1700002046.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700019494.mm.1 | NM_001115075.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700019494.mm.1 | NM_001115075.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700019494.mm.1 | TC1700002046.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700019493.mm.1 | NM_001115075.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700019493.mm.1 | NM_001115075.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700019493.mm.1 | TC1700002046.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700019498.mm.1 | NM_001115075.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700019498.mm.1 | NM_001115075.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700019498.mm.1 | TC1700002046.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700019490.mm.1 | NM_001115075.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700019490.mm.1 | NM_001115075.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700019490.mm.1 | TC1700002046.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700019496.mm.1 | NM_001115075.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700019496.mm.1 | NM_001115075.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700019496.mm.1 | TC1700002046.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 580121 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.1 expression beadchip | ILMN_1240555 | - | - |
