Assay Details
| Gene Symbol: Gm13630 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK034270.1 | - | 7 - 8 | 1993 | 91 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
sodium channel, voltage-gated, type I, alpha |
Scn1a |
B230332M13, Nav1.1 |
Anti-sense | Chr. 2 - 66270781 - 66440847 on NCBI Build GRCm38 | |
| Downstream | predicted gene 13629 | Gm13629 | OTTMUSG00000013098 | 19.3kb | Sense | Chr. 2 - 66440880 - 66547472 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm13630
Genomic Map