Assay Details
| Gene Symbol: Mep1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008585.2 | NP_032611.2 | 7 - 8 | 612 | 86 | |
| GenBank mRNA | AK093914.1 | - | 7 - 8 | 573 | 86 | |
| AK143906.1 | - | 7 - 8 | 570 | 86 | ||
| AK164734.1 | - | 7 - 8 | 567 | 86 | ||
| BC015258.1 | - | 7 - 8 | 612 | 4241252 | 86 | |
| M74897.1 | - | 7 - 8 | 611 | 86 | ||
More Information
Set Membership: |
|

Mep1a
Genomic Map