Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700007981.mm.1 |
NM_133776.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700007981.mm.1 |
NM_133776.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700007981.mm.1 |
TC1700000832.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700007977.mm.1 |
NM_133776.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700007977.mm.1 |
NM_133776.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700007977.mm.1 |
TC1700000832.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700007979.mm.1 |
NM_133776.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700007979.mm.1 |
NM_133776.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700007979.mm.1 |
TC1700000832.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700007976.mm.1 |
NM_133776.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700007976.mm.1 |
NM_133776.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700007976.mm.1 |
TC1700000832.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700007978.mm.1 |
NM_133776.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700007978.mm.1 |
NM_133776.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700007978.mm.1 |
TC1700000832.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700007980.mm.1 |
NM_133776.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700007980.mm.1 |
NM_133776.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700007980.mm.1 |
TC1700000832.mm.1 |
MTA TC |