Assay Details
| Gene Symbol: 4930543N07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016032.1 | - | 3 - 4 | 491 | 122 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transcription elongation regulator 1-like | Tcerg1l | 5730476P14Rik | -158.5kb | Anti-sense | Chr. 7 - 138208972 - 138397730 on NCBI Build GRCm38 |
| Downstream | mitogen-activated protein kinase 1 interacting protein 1 | Mapk1ip1 |
2310009E07Rik, AU043776, MISS |
271.9kb | Anti-sense | Chr. 7 - 138835818 - 138846267 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

4930543N07Rik
Genomic Map