Assay Details
| Gene Symbol: Jakmip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028708.2 | NP_082984.1 | 72 | 81 | ||
| GenBank mRNA | AK018189.2 | - | 74 | 81 | ||
| AK220482.1 | - | 52 | 81 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK018189.2, NM_028708.2 transcripts.
Set Membership: |
|

Jakmip3
Genomic Map