Assay Details
| Gene Symbol: Jakmip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028708.2 | NP_082984.1 | 311 | 72 | ||
| GenBank mRNA | AK018189.2 | - | 313 | 72 | ||
| AK220482.1 | - | 291 | 72 | |||
| BC119557.1 | - | 201 | 72 | |||
| BC119558.1 | - | 198 | 72 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Jakmip3
Genomic Map