Assay Details
| Gene Symbol: Gm5091 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_046164.1 | 1 - 2 | 87 | 69 | ||
| GenBank mRNA | AK082876.1 | - | 1 - 2 | 88 | 69 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ER membrane associated RNA degradation | Ermard |
2210404J11Rik, 2410011O22Rik, AI448938 |
-150.2kb | Anti-sense | Chr. 17 - 15041563 - 15090045 on NCBI Build GRCm38 |
| Downstream | predicted gene 10510 | Gm10510 | ENSMUSG00000073453 | 110.0kb | Sense | Chr. 17 - 15350813 - 15354461 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm5091
Genomic Map