Assay Details
| Gene Symbol: Gm9928 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK044473.1 | - | 2 - 3 | 524 | 86 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ceramide kinase-like | Cerkl |
Gm1958, Rp26 |
Anti-sense | Chr. 2 - 79331105 - 79430520 on NCBI Build GRCm38 | |
| Downstream | neurogenic differentiation 1 | Neurod1 |
BETA2, BHF-1, Neurod, bHLHa3 |
38.9kb | Anti-sense | Chr. 2 - 79452641 - 79456636 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm9928
Genomic Map