Assay Details
| Gene Symbol: 9430019J16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040635.1 | 2 - 3 | 263 | 127 | ||
| GenBank mRNA | AK020426.1 | - | 2 - 3 | 264 | 127 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 13652 | Gm13652 | OTTMUSG00000013241 | -297.7kb | Sense | Chr. 2 - 75126034 - 75138356 on NCBI Build GRCm38 |
| Downstream | heterogeneous nuclear ribonucleoprotein A3 | Hnrnpa3 |
2410013L13Rik, 2610209F03Rik, 2610510D13Rik, Hnrpa3 |
221.4kb | Anti-sense | Chr. 2 - 75659230 - 75669407 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 678389 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 284399 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P103946 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343980 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21344000 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01929949 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343873 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21344010 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343953 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343962 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01914335 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02233987 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343857 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343955 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02138332 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343921 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343913 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02138325 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343892 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01470028 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343952 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02233988 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343961 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343919 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01987532 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343947 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343927 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343882 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02138330 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343981 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343867 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343933 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343872 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343854 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343951 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343893 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343928 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02079942 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02138324 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02138339 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343945 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02118602 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343925 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01433949 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21344005 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343878 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02138323 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343834 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343994 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343881 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343978 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01406816 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02138327 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343999 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343838 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343971 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21344016 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02233980 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01872124 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02233983 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01844610 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343864 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343855 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02138335 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343958 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343908 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343983 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343884 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343917 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02061210 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343964 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343853 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343948 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343877 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02079951 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01780323 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343932 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343957 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343871 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343936 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343930 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343865 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21344011 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01433950 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343950 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01727027 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343956 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02138316 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343979 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343859 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01914336 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343886 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343968 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343840 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21344014 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02138334 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343923 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01388472 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343885 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01872122 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01727028 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01872129 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01303338 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02138337 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01727019 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01872125 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21343973 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Clariom S Array | PSR0200032383.mm.1 | NR_040635.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200032383.mm.1 | NR_040635.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200032383.mm.1 | TC0200003784.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200032385.mm.1 | NR_040635.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200032385.mm.1 | NR_040635.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200032385.mm.1 | TC0200003784.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200032384.mm.1 | NR_040635.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200032384.mm.1 | NR_040635.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200032384.mm.1 | TC0200003784.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 419437 | - | - |

9430019J16Rik
Genomic Map