Assay Details
| Gene Symbol: Gm14244 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK029760.1 | - | 2 - 3 | 1076 | 156 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
myelin and lymphocyte protein, T cell differentiation protein |
Mal |
Mpv17, Vip17 |
-36.7kb | Anti-sense | Chr. 2 - 127633226 - 127656695 on NCBI Build GRCm38 |
| Downstream |
mal, T cell differentiation protein-like |
Mall |
BC012256, BENE |
8.7kb | Anti-sense | Chr. 2 - 127704390 - 127729897 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm14244
Genomic Map